Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE1342

Sigma-Aldrich

Anti-ZBTB4 (KAISO-L1)

from rabbit

Synonym(s):

Zinc finger and BTB domain-containing protein 4, KAISO-like zinc finger protein 1, KAISO-L1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$475.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ZBTB4(57659)

General description

Zinc finger and BTB domain-containing protein 4 (UniProt: Q9P1Z0; also known as KAISO-like zinc finger protein 1, KAISO-L1, ZBTB4) is encoded by the ZBTB4 (also known as KIAA1538) gene (Gene ID: 57659) in human. ZBTB4 is an epigenetic regulator that localizes at pericentromeric heterochromatin. It is shown to be an essential component in maintaining genomic stability in mammals. It serves as a transcriptional repressor with bimodal DNA-binding specificity. It represses transcription in a methyl-CpG-dependent manner and binds with a higher affinity to methylated CpG dinucleotides in the consensus sequence 5′-CGCG-3′, but can also bind to the non-methylated consensus sequence 5′-CTGCNA-3′ (also known as the consensus kaiso binding site). ZBTB4 contains six C2H2-type zinc-finger domains and two proline-rich regions (aa 435-536 and 610-654). Its BTB (Broad-Complex, Tramtrack and Bric a brac) domain is localized in the amino acids 30-152. It is shown to be is frequently downregulated in cancer. ZBTB4 is phosphorylated by HIPK2 (Homeodomain Interacting Protein Kinase 2) and phosphorylation reduces its stability and triggers its degradation in response to DNA damage. (Ref.: Roussel-Gervais A., et al. (2017). Cancer Res; 77(1); 62-73; Yamada, D., et al (2009). Oncogene. 28(27); 2535-2544).

Specificity

This rabbit polyclonal antibody detects Zinc finger and BTB domain-containing protein 4 (KAISO-L1) in human cells. It targets an epitope within 17 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the internal region of human Zinc finger and BTB domain-containing protein 4.

Application

Anti-ZBTB4 (KAISO-L1), Cat. No. ABE1342, is a highly specific rabbit polyclonal antibody that targets Zinc finger and BTB domain-containing protein 4 and has been tested for use in Immunohistochemistry (Paraffin) and in Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected ZBTB4 (KAISO-L1) in human fallopian tube and human cervical cancer tissue sections.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected ZBTB4 (KAISO-L1) in PC3 cell lysate.

Target description

~105 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service