Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1081

Sigma-Aldrich

Anti-JMJD2E/KDM4E Antibody

serum, from rabbit

Synonym(s):

Lysine-specific demethylase 4E, KDM4D-like protein, Lysine-specific demethylase 4D-like, JMJD2E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$548.00

$548.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$548.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$548.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

NCBI accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM4E(390245)

General description

Histone N(ϵ)-methyl lysine demethylases are important in epigenetic regulation. JMJD2E (KDM4E) also known as Lysine-specific demethylase 4E, histone lysine demethylase 4E, or KDM4D-like protein, and encoded by the gene KDM4E/KDM4DL, is a member of the large Fe(II)/2-oxoglutarate- dependent family of human histone demethylases. JMJD2E (KDM4E) is a histone demethylase that specifically demethylates ′Lys-9′ of histone H3, thereby playing a central role in histone code. The KDM4 family also includes KDM4A, KDM4B, KDM4C, and KDM4D. These are also referred to as JMDM3A/JMJD2A, JMDM3B/JMJD2B, JMDM3C/JMJD2C, and JMDM3D/JMJD2D, respectively. Steady-state assays showed that KDM4E has a graded response to O2 over a physiologically relevant range of O2 concentrations, with activity level being dependent upon the methylation status of the target protein, and research with JMJD2E(KDM4E) may indicate the potential for histone demethylase activity to be regulated by oxygen availability.

Immunogen

Linear peptide corresponding to JMJD2E/KDM4E.

Application

Detect JMJD2E/KDM4E using this Anti-JMJD2E/KDM4E antibody validated for use in western blotting.

Quality

Evaluated by Western Blotting in PC-12 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected JMJD2E/KDM4E in 10 µg of PC-12 cell lysate.

Target description

~65 kDa observed

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingyi Li et al.
Nature communications, 13(1), 1172-1172 (2022-03-06)
Hypoxia is a physiological stress that frequently occurs in solid tissues. Autophagy, a ubiquitous degradation/recycling system in eukaryotic cells, renders cells tolerant to multiple stressors. However, the mechanisms underlying autophagy initiation upon hypoxia remains unclear. Here we show that protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service