Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1048

Sigma-Aldrich

Anti-GABPB-1/NRF2b Antibody

serum, from rabbit

Synonym(s):

GA-binding protein subunit beta-1, GABP subunit beta-1, GABPB-1, GABP subunit beta-2, GABPB-2, Nuclear respiratory factor 2, Transcription factor E4TF1-47, Transcription factor E4TF1-53, GABPB-1/NRF2b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on high sequence homology), equine (based on high sequence homology)

technique(s)

ChIP: suitable
electrophoretic mobility shift assay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GABPB2(126626)

General description

GA-binding protein subunit beta-1 (EC 3.2.1.3; UniProt Q06547; also known as GABP subunit beta-1, GABP subunit beta-2, GABPB-1, GABPB-2, Nuclear respiratory factor 2, Transcription factor E4TF1-47, Transcription factor E4TF1-53) is encoded by the GABPB1 (also known as E4TF1, E4TF1B, GABPB, GABPB2, NRF2B1, NRF2B2) gene (Gene ID 2553) in human. Initially identified through their interactions with cytochrome C and cytochrome oxidase promoters, GABP (NRF-2) and the related nuclear respiratory factor NRF-1 target many nuclear genes essential for mitochondrial respiratory function, including the mitochondrial transcription specificity factors TFB1M and TFB2M. GABP plays an anti-oxidant, pro-survival role in mitochondrial biogenesis process. GABP is a tetrameric complex composed of the NRF-2alpha (GABPA) and the NRF-2beta (GABPB) subunits. GABPA binds DNA via its ETS domain, and it recruits GABPB, which activates transcription through a glutamine-rich region in its carboxy terminus. GABPB and PRC, a member of PGC-1 family of regulated coactivators, can both interact with host cell factor 1 (HCF-1). PRC can therefore associate with GABP indirectly via HCF-1 and participate in GABP-dependent expression of the mitochondrial transcription factors TFB1M and TFB2M.

Specificity

Expected to react with all 4 spliced variants: isoform 1 (GABPB-1, Beta-1), isoform 2 (Beta-2), isoform 3 (GABPB-2, Gamma-1), isoform 4 (Gamma-2)

Immunogen

Recombinant protein corresponding to human GABPB-1/NRF2b.

Application

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected GABPB (NRF-2beta) occupancy at the TFB1M and TFB2M promoters by ChIP using human embryonic kidney 293FT chromatin preparations (Vercauteren, K., et al. (2008). J Biol Chem. 283(18):12102-12111).
Electrophoretic Mobility Shift Assay (EMSA): A representative lot caused a supershift of GABP A2/B2 (NRF-2 α2/β2) heterotetramer-DNA complex in EMSA using in vitro translated GABP (NRF-2) subunits and radiolabeled cytochrome oxidase subunit IV promoter fragment containing tandem GABP (NRF-2) recognition sites (Vercauteren, K., et al. (2008). J Biol Chem. 283(18):12102-12111).
Immunoprecipitation Analysis: A representative lot immunoprecipitated GABPB (NRF-2beta)-PRC complex from human embryonic kidney 293FT cell lysate (Vercauteren, K., et al. (2008). J Biol Chem. 283(18):12102-12111).
Western Blotting Analysis: A representative lot detected GABPB (NRF-2beta) in U2OS cell lysates (Gleyzer, N., and Scarpulla, R.C. (2011). J. Biol. Chem. 286(46):39715-39725).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-GABPB-1/NRF2b Antibody is validated for use in Western Blotting, Chromatin Immunoprecipitation (ChIP), Electrophoretic Mobility Shift Assay for the detection of GABPB-1/NRF2b.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected GABPB-1/NRF2b in 10 µg of HeLa cell lysate.

Target description

~48 kDa observed

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service