Skip to Content
MilliporeSigma
All Photos(3)

Documents

ABD75

Sigma-Aldrich

Anti-THAP11 (Ronin) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

THAP domain-containing protein 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... THAP11(57215)

General description

THAP11 (Ronin) is a transcriptional repressor which is predominantly expressed in embryonic stem (ES) cells. It contains a THAP-type zinc finger domain which is characteristic of the THAP 11 family members. In ES cells, THAP11 interacts with the transcriptional co-regulator HCF-1, to recruit chromatin-modifying proteins such as members of the polycomb family of proteins, to induce gene repression. THAP11 targets genes involved in metabolism, transcriptional initiation, and messenger RNA splicing. It plays an important role in maintaining the pluripotent state of ES cells.

Immunogen

KLH-conjugated linear peptide corresponding to human THAP11.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-THAP11 (Ronin) Antibody is validated for use in Western Blotting, IHC for the detection of THAP11 (Ronin).
Western Blot Analysis: 1 µg/mL from a representative lot detected THAP11 on 10 µg of mouse liver tissue lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected THAP11 in mouse cardiac muscle and mouse liver tissues.

Quality

Evaluated by Western Blot in mouse heart tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected THAP11 on 10 µg of mouse heart tissue lysate.

Target description

~48 kDa observed. Uncharacterized bands appear at ~20-25 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse heart tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service