Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABD136

Sigma-Aldrich

Anti-LGI1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-rich glioma inactivated protein 1, Epitempin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LGI1(9211)

General description

Leucine-rich glioma inactivated gene 1 (LGI1), also known as Epitempin-1 (EPT), is a member of a family of highly related proteins containing leucine-rich repeats (LRRs) regulating voltage-gated potassium channels assembled from KCNA1, KCNA4 and KCNAB1. LGI1 may play a role in the control of neuroblastoma cell survival and may play a role in other cancers. It is expressed in neural tissue, especially brain. Mutations in LGI1 have been associated with nearly half of autodominant lateral temporal epilepsy (ADTLE).

Specificity

This antibody recognizes the N-terminus of LGI1.

Immunogen

KLH-conjugated linear peptide corresponding to the N-terminus of human LGI1.

Application

Detect LGI1 using this rabbit polyclonal antibody, Anti-LGI1 Antibody validated for use in western blotting.

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in mouse brain tissue lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 1 µg/mL of this antibody detected LGI1 in 15 µg of mouse brain tissue lysate.

Target description

~56 kDa observed. Uniprot describes 3 isoforms at ~33 kDa, ~58 kDa, and ~64 kDa Uncharacterized bands may appear in some lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service