Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABC845

Sigma-Aldrich

Anti-Nop30 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nucleolar protein 3, Apoptosis repressor with CARD, Muscle-enriched cytoplasmic protein, Myp, Nucleolar protein of 30 kDa, Nop30

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOL3(8996)

General description

Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Nop30 (also known as Nucleolar protein 3, Apoptosis repressor with CARD, Muscle-enriched cytoplasmic protein, Myp, Nucleolar protein of 30 kDa, and Nop30) is an alternate splicing isoform of ARC. While ARC interacts with caspase-2 and –8 and suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR-1 and DR3, Nop30 multimerizes and binds to the splicing factor SRp30c and may act to influence alternative splice site selection in vivo.

Specificity

This Nop30 antibody will not detect ARC protein.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to a 14 amino acid peptide near the C-terminus of human Nop30.

Application

Anti-Nop30 Antibody is a highly specific rabbit polyclonal antibody, that targets Nucleolar protein 3 & has been tested in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in mouse muscle tissue lysate.

Western Blotting Analysis: 2 μg/mL of this antibody detected Nop30 in mouse muscle tissue lysate.

Target description

~24 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service