Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC832

Sigma-Aldrich

Anti-TMEM16B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Anoctamin-2, Transmembrane protein 16B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANO2(57101)

General description

Anoctamin-2, also known as Transmembrane protein 16B (TMEM16B) are subunits of Calcium-activated chloride channels (CaCC) which mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. TMEM16B is predicted to have eight transmembrane domains with both the amino and carboxy termini in the cytoplasm and is expressed in several tissues including olfactory sensory neurons as well as photoreceptors in mammalian retina. Like TMEM16A, TMEM16B is thought to form at least part of CaCC’s but has different biophysical characteristics such as voltage dependence and unitary conductance.

Specificity

This antibody will not cross-react with TMEM16A.

Immunogen

Epitope: N-terminus
KLH-conjugated synthetic peptide corresponding to the N-terminus of human TMEM16B.

Application

Anti-TMEM16B Antibody is an antibody against TMEM16B for use in western blotting, IHC & immunofluorescence.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected TMEM16B in human brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected TMEM16B in human brain cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in rat brain tissue.

Western Blotting Analysis: 2 μg/mL of this antibody detected TMEM16B in rat brain tissue.

Target description

~128 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service