Skip to Content
MilliporeSigma
All Photos(2)

Documents

ABC506

Sigma-Aldrich

Anti-FUNDC1 Antibody

serum, from rabbit

Synonym(s):

FUN14 domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FUNDC1(139341)

General description

FUN14 domain-containing protein 1 (FUNDC1) is an important autophagy partner for mitophagy, the process whereby mitochondria get recycled after damage. FUNDC1 is localized to the outer mitochondrial membrane and widely expressed in all cells and appears to act as an activator of hypoxia-induced mitophagy. FUNDC1 interacts directly with important autophagy markers such as MAP1LC3B and GABARAP. Phosphorylation at Tyr-18 inactivates FUNDC1 mitophagy activation ability, and it is this dephosphorylation following hypoxia that initiates the interaction with MAP1LC3 family proteins and mitophagy. Recent research shows that FUNDC1 expression is controlled via the microRNA miRNA-137, and that this microRNA is critical for mitophagy and mitochondrial recycling.

Immunogen

Epitope: Sigma transmembrane domain
Recombinant protein corresponding to the Sigma transmembrane domain of Human FUNDC1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-FUNDC1 Antibody is validated for use in Western Blotting and Immunocytochemistry and Immunoprecipitation for the detection of FUNDC1.
Western Blotting Analysis: A representatitive lot of this antibody was used to detect FUNDC1 in A431, C2C12, HEK293, HeLa, HepG2, L6, Mouse brain, NIH/3T3, PC12, PC3, and RAW264.7 cell lysates. This antibody did not detect FUNDC1 in Human placenta, HUVEC, or Jurkat cell lysates.
Western Blotting Analysis: A representatitive lot of this antibody was used to detect puried FUNDC1 protein from bacteria or HeLa cell lysates (Liu, L., et al. (2012) Nature Cell Biology. 14(2):177-185).
Immunocytochemistry Analysis: A representatitive lot of this antibody was used to detect FUNDC1 in HeLa cells (Liu, L., et al. (2012) Nature Cell Biology. 14(2):177-185.
Immunoprecipitation Analysis: A representatitive lot of this antibody was used to co-immunoprecipitate FUNDC1 with endogenous LC3 in HeLa cells that were exposed to normoxic or hypoxic conditions for 24 hrs (Liu, L., et al. (2012) Nature Cell Biology. 14(2):177-185).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected FUNDC1 in 10 µg of HepG2 cell lysate.

Target description

~17 kDa observed.
Uncharacterized bands may be visitible in some lysates.

Physical form

Rabbit Polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Wang et al.
Nature communications, 12(1), 2616-2616 (2021-05-12)
FUN14 domain-containing protein 1 (FUNDC1) is an integral mitochondrial outer-membrane protein, and mediates the formation of mitochondria-associated endoplasmic reticulum membranes (MAMs). This study aims to determine the contributions of FUNDC1-mediated MAMs to angiogenesis in vitro and in vivo. In cultured
Wen-Qi Ma et al.
Cell death & disease, 11(11), 991-991 (2020-11-19)
Pyruvate dehydrogenase kinase 4 (PDK4) is an important mitochondrial matrix enzyme in cellular energy regulation. Previous studies suggested that PDK4 is increased in the calcified vessels of patients with atherosclerosis and is closely associated with mitochondrial function, but the precise
Mingjie Yang et al.
British journal of pharmacology, 178(4), 964-982 (2020-12-11)
Chronic alcohol consumption contributes to contractile dysfunction and unfavourable geometric changes in myocardium, accompanied by altered autophagy and disturbed mitochondrial homeostasis. The E3 ubiquitin ligase Parkin encoded by PARK2 gene maintains a fundamental role in regulating mitophagy and mitochondrial homeostasis
Panchanan Maiti et al.
International journal of molecular sciences, 20(2) (2019-01-24)
Autophagy and the (PI3K-Akt/mTOR) signaling pathway play significant roles in glioblastoma multiforme (GBM) cell death and survival. Curcumin (Cur) has been reported to prevent several cancers, including GBM. However, the poor solubility and limited bioavailability of natural Cur limits its
Kibrom M Alula et al.
Scientific reports, 13(1), 18-18 (2023-01-03)
Autophagy of damaged mitochondria, called mitophagy, is an important organelle quality control process involved in the pathogenesis of inflammation, cancer, aging, and age-associated diseases. Many of these disorders are associated with altered expression of the inner mitochondrial membrane (IMM) protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service