Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABC460

Sigma-Aldrich

Anti-Sec23B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Tumor-associated calcium signal transducer 2, Cell surface glycoprotein Trop-2, Membrane component chromosome 1 surface marker 1, Pancreatic carcinoma marker protein GA733-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TACSTD2(4070)

General description

The protein named Sec23B, also known as protein transport protein Sec23B, or SEC23-related protein B is a component of the COPII coat, the complex that governs ER-derived vesicles involved in transport from the endoplasmic reticulum to the Golgi apparatus. The transport of proteins in eukaryotic cells from the endoplasmic reticulum (ER) to the Golgi complex proceeds by deformation of specialized portions of the donor membrane to form carrier vesicles. A group of cytosolic proteins collectively known as COPII carry out a programmed set of sequential interactions, leading to cargo sorting and vesicle budding. The Sec23B protein is critical to proper protein transport in and out of cells, and it is localized to the smooth endoplasmic reticulum membrane where it is a peripheral membrane protein and interacts with the Golgi apparatus membranes as well. Sec23B is expressed at high levels in brain and fibroblasts as well as liver and other metabolically active tissues. Defects in Sec23B cause congenital dyserythropoietic anemia type II, a recessive disorder of erythroid differentiation but the disease is not caused by loss of function of Sec23B but rather a combination of a hypomorphic allele with altered functionality appears to be the cause and its related protein Sec23A can often compensate somewhat for the defect.

Immunogen

Linear peptide corresponding to mouse Sec23B.

Application

Detect Sec23B using this rabbit polyclonal antibody, Anti-Sec23B Antibody validated for use in western blotting.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Western Blotting Analysis: A representative lot from an independent laboratory detected Sec23B in various mouse tissue lysates expressing wild-type Sec23B. The same lot detected Sec23B in Sec23B wild-type MEF cell lysates and demonstrated a loss of signal in Sec23B knockdown MEF cell lysates (Tao, J., et al. (2012). Proc Natl Acad Sci USA. 109(29):E2001-E2009.).

Quality

Evaluated by Western Blotting in mouse pancreas tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Sec23B in 10 µg of mouse pancreas tissue lysate.

Target description

~86 kDa observed. Uncharacterized Band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Wei et al.
The Journal of biological chemistry, 298(1), 101536-101536 (2021-12-27)
SEC23B is one of two vertebrate paralogs of SEC23, a key component of the coat protein complex II vesicles. Complete deficiency of SEC23B in mice leads to perinatal death caused by massive degeneration of professional secretory tissues. However, functions of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service