Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC259

Sigma-Aldrich

Anti-Death-associated protein 1 Antibody

1.0 mg/mL, from rabbit

Synonym(s):

Death-associated protein 1, DAP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
human, mouse
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DAP(1611)

General description

The protein Death-associated protein 1 or also known as DAP-1/DAP1 and encoded by the gene DAP/DAP1 is a negative regulator of autophagy. The link of DAP1 to autophagy was recognized because in DAP1 knockdowns there was enhanced autophagic flux and DAP1 displayed a rapid decline in its phosphorylation in response to amino acid starvation, a key inducer of autophagy. Mapping of the phosphorylation sites and analysis of phosphorylation mutants indicated that DAP1 is functionally silenced in growing cells through mTOR-dependent phosphorylation on Ser3 and Ser51. Inactivation of mTOR during starvation caused a rapid reduction in these phosphorylation sites and converted the protein into an active suppressor of autophagy. These results are consistent with a "Gas and Brake" model in which mTOR inhibition also controls a buffering mechanism that counterbalances the autophagic flux and prevents its over activation under nutrient deprivation. DAP1 is also involved in mediating interferon gamma induced cell death, which appears to often be mediated by autophagy, and recently it has been shown to be important in regulating myogenesis and the apoptosis of satellite cells during development. DAP1/Death-associated protein 1 is regulated by phosphorylation; phosphorylation by MTOR inhibits the suppressive activity of DAP toward autophagy. DAP1 is expressed by most cells and up regulated in many cancer cells as well.

Immunogen

KLH-conjugated linear peptide corresponding to the unknown of human Death-associated protein 1.

Application

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Death-associated protein 1 in human prostate cancer and normal testis tissues. (Note: It is recommended for end-users to perform a 1:1,000 dilution for enhanced performance.)
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Death-associated protein 1 Antibody is validated for use in western blotting & IHC for the detection of Death-associated protein 1.

Quality

Evaluated by Western Blotting in serum starved and control NIH/3T3 cell lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected Death-associated protein 1 in serum starved NIH/3T3 cell lysate and demonstrated a loss of signal in control NIH/3T3 cell lysate.

Target description

~15 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service