Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC227

Sigma-Aldrich

Anti-GABARAPL2

from rabbit, purified by affinity chromatography

Synonym(s):

Gamma-aminobutyric acid receptor-associated protein-like 2, GABA(A) receptor-associated protein-like 2, Ganglioside expression factor 2, GEF-2, General protein transport factor p16, Golgi-associated ATPase enhancer of 16 kDa, GATE-16, MAP1 light chain 3-

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Gamma-aminobutyric acid receptor-associated protein-like 2 (UniProt: P60520; also known as GABA(A) receptor-associated protein-like 2, Ganglioside expression factor 2, GEF-2, General protein transport factor p16, Golgi-associated ATPase enhancer of 16 kDa, GATE-16, MAP1 light chain 3-related protein) is encoded by the GABARAPL2 (also known as FLC3A, GEF2) gene (Gene ID: 11345) in human. GEF-2 is a momomeric ubiquitin-like modifier protein of ATG8 family that is involved in intra-Golgi traffic. It is shown to modulate intra-Golgi transport through coupling between NSF activity and SNAREs activation. It first stimulates the ATPase activity of NSF, which in turn stimulates the association with GOSR1. GEF-2 is also involved in autophagy and plays a role in mitophagy, which contributes to regulate mitochondrial quantity and quality by eliminating the mitochondria to a basal level. It is ubiquitously expressed and higher expressions are observed in the brain, heart, prostate, ovary, spleen and skeletal muscle. Low expression levels are observed in lung, thymus, and small intestine. In the precursor molecule the peptide bond between aa 116 and 117 is cleaved off by ATG4 to generate mature protein.

Specificity

This rabbit polyclonal antibody detects GABA(A) receptor-associated protein-like 2 (GABARAPL2) in human cells.

Immunogen

GST-tagged full length human recombinant GABA(A) receptor-associated protein-like 2 (GABARAPL2).

Application

Anti-GABARAPL2, Cat. No. ABC227, is a rabbit polyclonal antibody that detects Gamma-aminobutyric acid receptor-associated protein-like 2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected GABARAPL2 in human cerebellum and human prostate tissue sections.

Quality

Evaluated by Western Blotting in Raji membrane extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected GABARAPL2 in Raji membrane extract.

Target description

~ 14 kDa observed; 13.67 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide/.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service