Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABC1428

Sigma-Aldrich

Anti-RecQ1

from rabbit

Synonym(s):

ATP-dependent DNA helicase Q1, RecQ-like type 1, DNA-dependent ATPase Q1, RecQ protein-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RECQL(5965)

General description

ATP-dependent DNA helicase Q1 (UniProt: P46063; also known as EC:3.6.4.12, DNA helicase, RecQ-like type 1, RecQ1, DNA-dependent ATPase Q1, RecQ protein-like 1) is encoded by the RECQL (also known as RECQ1, RECQL1) gene (Gene ID: 5965) in human. RecQ helicases are a ubiquitous family of DNA unwinding enzymes involved in the maintenance of chromosome stability. They participate in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Five members of the RecQ family have been reported in human cells: BLM, RECQ1, RECQ4, RECQ5, and WRN. RecQ1 displays high expression in heart, lung, skeletal muscle and kidney. It is also highly expressed in tumor cells and although brain exhibits low level of EecQ1, its expression is dramatically increased in human glioblastoma. Depletion of RecQ1 affects proliferation of glioblastoma cells and causes an increase in the level of DNA damage. RecQ1 depleted tumor cells are shown to be more sensitive to anti-tumor agents, such as hydroxyurea and temozolomide.

Specificity

This rabbit polyclonal antibody detects ATP-dependent DNA helicase Q1 (RecQ1) in human cells. It targets an epitope within 16 amino acids from the C-terminal end.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human ATP-dependent DNA helicase Q1 (RecQ1).

Application

Anti-RecQ1, Cat. No. ABC1428, is a rabbit polyclonal antibody that detects ATP-dependent DNA helicase Q1 and has been tested for use in Immunofluorescence, Immunohistochemistry, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected RecQ1 in T98G cells after treatment with anti-RECQ1 siRNA or control siRNAs (Mendoza-Maldonado, R., et. al. (2011). Mol Cancer. 10:83).

Western Blotting Analysis: A representative lot detected RecQ1 in Western Blotting applications (Thangavel, S., et. al. (2015). J Cell Biol. 208(5):545-62).

Immunohistochemistry Analysis: A representative lot detected RecQ1 in tissues are perilesional sections from multiform glioblastoma, colon carcinoma,lung cancer and thyroid cancer (Mendoza-Maldonado, R., et. al. (2011). Mol Cancer. 10:83).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in IMR90 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected RecQ1 in 10 µg of IMR90 human fetal lung cell lysate.

Target description

~75 kDa observed; 73.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing Tris-citrate-phosphate buffer (pH 7.8) with 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonas Andreas Schmid et al.
Molecular cell, 71(6), 897-910 (2018-08-21)
Chromatin ubiquitination by the ubiquitin ligase RNF168 is critical to regulate the DNA damage response (DDR). DDR deficiencies lead to cancer-prone syndromes, but whether this reflects DNA repair defects is still elusive. We identified key factors of the RNF168 pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service