Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABC1389

Sigma-Aldrich

Anti-DCLK1-S

from rabbit

Synonym(s):

Serine/threonine-protein kinase DCLK1-S, EC: 2.7.11.1, Doublecortin domain-containing protein 3A-short, Doublecortin-like and CAM kinase-like 1, Doublecortin-like kinase 1, PS41014

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DCLK1(9201)

Related Categories

General description

Serine/threonine-protein kinase DCLK1 (UniProt: O15075; also known as EC: 2.7.11.1, Doublecortin domain-containing protein 3A, Doublecortin-like and CAM kinase-like 1, Doublecortin-like kinase 1) is encoded by the DCLK1 (also known as DCAMKL1, DCDC3A, KIAA0369) gene (Gene ID: 9201) in human. DCLK1 is an integral component of plasma membrane and is highly expressed in fetal brain tissue and is also detectable in lung and liver. DCLK1 expression is reported to be critical for growth of human colon cancer cells and its expression is up-regulated in response to growth factors resulting in hyperproliferation. Both long (L) (~80-82 kDa) and short (S) (~45-50 kDa) isoforms of DCLK1 have been described. The long isoform of DCLK1 contains two N-terminal doublecortin domains, which bind microtubules, a C-terminal serine/threonine kinase domain with homology to CaM kinases, and a middle serine/proline rich domain that mediates protein interactions. The short isoform, DCLK1-S, has been reported in human colorectal cancer cells and in cancer stem cells, while normal colon cells express only the long isoform of DCLK1. The short isoforms lacks the two N-terminal doublecortin domains. Although DCLK1-S differs from the long isoform by only six amino-acids., this mono-specific antibody does not cross-react with the long isoform and is shown to specifically detect cancer stem cells. The short isoform is not a splice variant, but is expressed from an alternate beta promoter in intron V of the gene that results in the presence of the unique sequence of amino acids. These amino acids are absent in the long isoform, which is expressed by the alpha promoter in the 5’ region of the gene. It has been reported that the 5′ (alpha)-promoter of the human DCLK1 gene becomes epigenetically silenced during colon carcinogenesis, resulting in loss of expression of the canonical long isoform in human colorectal adenocarcinomas (hCRCs). Instead, hCRCs express the short isoform, DCLK1-S. Colon tumors and normal colons from mice only express the long isoform, since the epigenetics of colon carcinogenesis in mice is significantly different from that in humans.(Ref.:Sarkar,S et al.(2017).Lab.Invest.97(10):1245-1261;Sarkar,S.,et al.(2017).Mol.Cancer Res.15(12):1678-1691).

Specificity

This rabbit polyclonal antibody detects the short version of Serine/threonine-protein kinase DCLK1 (DCLK1-S). It does not react with long isoforms. It targets an epitope within six amino acids from the N-terminus region.

Immunogen

A multimeric scaffold of the 6 amino acids from the N-terminal region of the short version of human DCLK1 (DCLK1-S).

Application

Anti-DCLK1-S, Cat. No. ABC1389, is a rabbit polyclonal antibody that specifically detects the short version of Serine/threonine-protein kinase DCLK1. It has been tested for use in Immunohistochemistry and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected DCLK1-S in Immunohistochemistry applications (Sarkar, S., et. al. (2017). Lab Invest. [Epub ahead of print]).

Western Blotting Analysis: A representative lot detected DCLK1-S in Western Blotting applications (Sarkar, S., et. al. (2017). Lab Invest. [Epub ahead of print]).

Quality

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected DCLK1-S in 10 µg of HCT116 cell lysate.

Target description

~55 kDa observed. 47.60 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service