Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9690

Sigma-Aldrich

Anti-NMDAR2B phospho Tyrosine 1336 Antibody

Chemicon®, from rabbit

Synonym(s):

NR2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1336)

Gene Information

Specificity

NMDAR2B, phosphoTyrosine1336. The antibody recognizes a protein of ~180 kDa corresponding to NMDAR2B, phosphoTyrosine1336 in lysates from rat brain hippocampus. Immunolabeling is blocked by the phosphopeptide used as the immunogen but not the corresponding non-phosphopeptide. Immunolabeling is also blocked by l-phosphatase treatment.

Immunogen

Synthetic peptide of amino acids surrounding the phosphoTyrosine 1336 site of rat NMDAR2B. The immunogen has 100% similarity with human and mouse.

Application

Detect NMDAR2B phospho Tyrosine 1336 using this Anti-NMDAR2B phospho Tyrosine 1336 Antibody validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:1,000.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: 07-533

Physical form

Affinity purified immunoglobulin. Liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérémy Ferrier et al.
PloS one, 8(10), e77828-e77828 (2013-11-10)
Oxaliplatin is an anticancer drug used for the treatment of advanced colorectal cancer, but it can also cause painful peripheral neuropathies. The pathophysiology of these neuropathies has not been yet fully elucidated, but may involve spinal N-methyl-D-aspartate (NMDA) receptors, particularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service