Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9303

Sigma-Aldrich

Anti-Trace Amine Receptor 1 Antibody

Chemicon®, from rabbit

Synonym(s):

TA1, TAAR1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$537.00

$537.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$537.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$537.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TAAR1(134864)

Specificity

Recognizes Trace Amine Receptor 1 [TAAR1, TA1].
The immunogen has 63% similarity with mouse.

Immunogen

Synthetic peptide from the 3rd cytoplasmic domain of human Trace Amine Receptor 1.

Application

Anti-Trace Amine Receptor 1 Antibody detects level of Trace Amine Receptor 1 & has been published & validated for use in IH(P) & WB.
Immunohistochemistry on paraffin embedded tissue sections.

Western blot.

Optimal working dilutions must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service