Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9074

Sigma-Aldrich

Anti-IP3 Receptor 2 Antibody

serum, Chemicon®

Synonym(s):

Inositol 1,4,5-Trisphosphate Receptor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$622.00

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$622.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITPR2(3709)

Specificity

IP3 Receptor 2.

Immunogen

Synthetic peptide from the C terminus of human IP3 Receptor 2.

Application

Detect IP3 Receptor 2 using this Anti-IP3 Receptor 2 Antibody validated for use in IC, IH & WB.
Research Category
Signaling
Research Sub Category
Lipid Signaling
Western blot: 1:1,000 using ECL. The antibody reacts with the 250 kDa IP3 Receptor 2 protein in microsomal preparations. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Preferred gel percentage is 4-12% gradient gel or 5% PAGE. For microsomal methods and blotting procedure similar antibodies can be found in: Perez, et al. J Biol. Chem. 272:23961

http://www.jbc.org/cgi/reprint/272/38/23961?ijkey=35bcf508a3025fc8bf8203ea3805122eec0caf1e )

Immunocytochemistry: 1:1,000

Immunohistochemistry: 1:1,000 overnight at 2-8°C using a fluorescently labeled secondary antibody. Suggested fixative is 4% paraformaldehyde in 0.1M PBS (one hour). Suggested permeablization method is 0.05% Triton X-100 in dilution buffer. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS.

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Western blot = adult mouse liver.

IHC = adult mouse hepatocytes (adult mouse cerebellum Purkinje cells will be negative).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-yeon Hwang et al.
Cell calcium, 45(3), 233-242 (2008-12-17)
Progesterone (P4) is a steroid hormone that plays multiple roles in the central nervous system (CNS) including promoting neuroprotection. However, the precise mechanisms involved in its neuroprotective effects are still unknown. Given that the regulation of the intracellular calcium (Ca(2+))
Function and expression of ryanodine receptors and inositol 1,4,5-trisphosphate receptors in smooth muscle cells of murine feed arteries and arterioles.
Westcott, EB; Goodwin, EL; Segal, SS; Jackson, WF
The Journal of Physiology null
S Kaja et al.
Neuroscience, 175, 281-291 (2010-11-16)
Dysregulation of Ca(2+) signaling following oxidative stress is an important pathophysiological mechanism of many chronic neurodegenerative disorders, including Alzheimer's disease, age-related macular degeneration, glaucomatous and diabetic retinopathies. However, the underlying mechanisms of disturbed intracellular Ca(2+) signaling remain largely unknown. We
Calcium Receptor and Nitric Oxide Synthase Expression in Circular Muscle of Lower Esophagus from Patients with Achalasia.
Yang Gao et al.
Chinese medical journal, 131(23), 2882-2885 (2018-12-05)
W E Medina-Ortiz et al.
The open ophthalmology journal, 1, 8-16 (2007-01-01)
We have determined the presence and cellular distribution of intracellular calcium channels, inositol 1, 4, 5-trisphosphate receptors (IP3Rs) and ryanodine receptors (RyRs) in adult and postnatal (P10) lacrimal gland acinar cells. Western blot analysis of both P10 cultures and adult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service