Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5856

Sigma-Aldrich

Anti-VAMP-2 Antibody

Chemicon®, from rabbit

Synonym(s):

Vesicle-Associated Membrane Protein 2, Synaptobrevin 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VAMP2(6844)

Specificity

Recognizes VAMP-2 (Vesicle-Associated Membrane Protein 2, Synaptobrevin 2). The epitope does not share homology with any other known proteins.

SPECIES REACTIVITIES: It is expected that the antibody will also react with human, Macaca multata and bovine due to sequence homology (25/26). Other species have not been tested.

Immunogen

GST fusion protein corresponding to a cytoplasmic, N-terminal part of rat or mouse VAMP-2 (Accession numbers Q64357, NP 036795).

Application

Anti-VAMP-2 Antibody detects level of VAMP-2 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot: 1:1,000 using ECL on rat brain membranes.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min)

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 50 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of PBS. For positive control, in Western blot using 20 ng of protein per Minigel lane. For negative control, preincubate 3 μg of fusion protein with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-François Bouchard et al.
Journal of neurochemistry, 107(2), 398-417 (2008-08-12)
The netrin-1 receptor Deleted in Colorectal Cancer (DCC) is required for the formation of major axonal projections by embryonic cortical neurons, including the corpus callosum, hippocampal commissure, and cortico-thalamic tracts. The presentation of DCC by axonal growth cones is tightly
Kwok-Ho Lam et al.
Cell reports, 30(8), 2526-2539 (2020-02-27)
Botulinum neurotoxin (BoNT) is one of the most acutely lethal toxins known to humans, and effective treatment for BoNT intoxication is urgently needed. Single-domain antibodies (VHH) have been examined as a countermeasure for BoNT because of their high stability and
Helmut Kubista et al.
Journal of cell science, 117(Pt 6), 955-966 (2004-02-06)
The core complex, formed by the SNARE proteins synaptobrevin 2, syntaxin 1 and SNAP-25, is an important component of the synaptic fusion machinery and shows remarkable in vitro stability, as exemplified by its SDS-resistance. In western blots, antibodies against one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service