Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB5808

Sigma-Aldrich

Anti-HCN4 Antibody

Chemicon®, from rabbit

Synonym(s):

Hyperpolarization-Activated Cyclic Nucleotide-Gated Potassium Channel 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HCN4(10021)

Specificity

Recognizes HCN4 (Hyperpolarization-Activated Cyclic Nucleotide-Gated Potassium Channel 4). The epitope does not share homology with any other known proteins.

Immunogen

GST fusion protein corresponding to amino acids 119-155 of human HCN4 (Accession XP_007730, NP_005468, CAB42604, CAB52754).

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-HCN4 Antibody is validated for use in IH, WB for the detection of HCN4.
Western blot: 1:200 using ECL on rat brain membranes.

Immunohistochemistry using rat brain sections.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 0.05% sodium azide as a preservative. Reconstitute with 200 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 120 μg of control antigen. The stock solution of the antigen can be made up using 100 μL of PBS. For positive control, in Western blot using 20 ng of protein per Minigel lane. For negative control, preincubate 3 μg of fusion protein with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanako Oi et al.
Visual neuroscience, 25(1), 95-102 (2008-02-20)
Antisera directed against hyperpolarization-activated, cyclic nucleotide-sensitive (HCN) channels bind to somata in the ganglion cell layer of rat and rabbit retinas, and mRNA for different HCN channel isoforms has been detected in the ganglion cell layer of mouse retina. However
Alicia D'Souza et al.
Nature communications, 5, 3775-3775 (2014-05-16)
Endurance athletes exhibit sinus bradycardia, that is a slow resting heart rate, associated with a higher incidence of sinus node (pacemaker) disease and electronic pacemaker implantation. Here we show that training-induced bradycardia is not a consequence of changes in the
Xian-zhi He et al.
Journal of cardiology, 60(3), 242-247 (2012-06-19)
This study aimed to discover the pathogenesis of focal atrial fibrillation (AF) originating from pulmonary veins by observing the histological structure and special cells in the canine pulmonary vein model of persistent atrial fibrillation. The pulmonary veins and the sinus
C A Remme et al.
Basic research in cardiology, 104(5), 511-522 (2009-03-04)
Cardiac sodium channels are responsible for conduction in the normal and diseased heart. We aimed to investigate regional and transmural distribution of sodium channel expression and function in the myocardium. Sodium channel Scn5a mRNA and Na(v)1.5 protein distribution was investigated
Gloria J Partida et al.
Investigative ophthalmology & visual science, 53(3), 1696-1703 (2012-01-28)
The membrane expression and gene promoter of the glycosylphosphatidylinositol (GPI)-anchored protein Thy1 have been widely used to examine the morphology and distribution of retinal ganglion cells in normal eyes and disease models. However, it is not known how adult mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service