Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB5246

Sigma-Aldrich

Anti-P2X7 Receptor Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... P2RX7(5027)

Specificity

The epitope is specific for P2X7 and is not present in any other known protein.

SPECIES REACTIVITIES: Rat, mouse (Solee et al. 2001) and Candida albicans (Koshlukova et al. 2000). The sequence is highly conserved in mouse and human (18/20 and 16/20 residues identical, respectively). Reactivity with other species has not been tested.

Immunogen

Purified peptide P2X7 from 576-595 of rat P2X7 (Koshlukova et al. 2000).

Application

Anti-P2X7 Receptor Antibody detects level of P2X7 Receptor & has been published & validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot:1:200-1:1000 using ECL on rat brain membranes.

Immunohistochemistry (Vial & Evans 2000; Lewis & Evans 2000; Gitterman & Evans 2000)

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 200 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile distilled water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan R Genzen et al.
Purinergic signalling, 5(3), 299-307 (2009-03-11)
Ependymal cells line the cerebral ventricles and are located in an ideal position to detect central nervous system injury and inflammation. The signaling mechanisms of ependymal cells, however, are poorly understood. As extracellular adenosine 5'-triphosphate is elevated in the context
Maria Pina Notarangelo et al.
Frontiers in cell and developmental biology, 12, 1368318-1368318 (2024-04-19)
A comprehensive understanding of the molecules that play key roles in the physiological and pathological homeostasis of the human intervertebral disc (IVD) remains challenging, as does the development of new therapeutic treatments. We recently found a positive correlation between IVD
Paola Chiozzi et al.
Scientific reports, 9(1), 6475-6475 (2019-04-26)
Previous data from our laboratory show that expression of the P2X7 receptor (P2X7R) is needed for amyloid β (Aβ)-stimulated microglia activation and IL-1β release in vitro and in vivo. We also showed that Aβ-dependent stimulation is inhibited by the dihydropyridine
Anna Lisa Giuliani et al.
Frontiers in immunology, 10, 793-793 (2019-04-30)
The P2X7 receptor (P2X7R) is a key pro-inflammatory plasma membrane receptor responsible for NLRP3 inflammasome activation and IL-1β release. Various inflammatory plasma membrane receptors (e.g., IL-1 type I receptor, TNF type I and II receptors, IL-2 receptor) are shed under
Ana A Q A Santos et al.
World journal of gastroenterology, 28(30), 4075-4088 (2022-09-27)
Clostridioides difficile (C. difficile) is the most common pathogen causing health care-associated infections. C. difficile TcdA and TcdB have been shown to activate enteric neurons; however, what population of these cells is more profoundly influenced and the mechanism underlying these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service