Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5050P

Sigma-Aldrich

Anti-NMDAR1 Splice Variant C2 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Grin1(14810)
rat ... Grin1(24408)

Related Categories

Specificity

Specific for the NMDAR1 containing the C2′ splice variant insert. The antibody does not recognize the NMDAR1 subunits of the NMDA receptor that do not contain the C2′ insert. By Western blot the antibody recognizes a protein with molecular weight of 120 kDa. Also labels a single higher molecular weight band that may be a posttranslationally modified form of the protein.

Immunogen

Peptide corresponding to the C-terminus of the rat NMDAR1 C2′ splice variant.

Application

Detect NMDAR1 Splice Variant C2 using this Anti-NMDAR1 Splice Variant C2 Antibody validated for use in IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:2000 using ECL

Immunohistochemistry: 1:1000

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from ammonium bicarbonate (5 mM) so some residual salt may also be present. Reconstitute with 50 μL of PBS. Contains no preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Withdrawal from chronic intermittent alcohol exposure increases dendritic spine density in the lateral orbitofrontal cortex of mice.
McGuier, NS; Padula, AE; Lopez, MF; Woodward, JJ; Mulholland, PJ
Alcohol null
Intrauterine growth restriction due to uteroplacental insufficiency decreased white matter and altered NMDAR subunit composition in juvenile rat hippocampi.
Schober, ME; McKnight, RA; Yu, X; Callaway, CW; Ke, X; Lane, RH
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology null
Thorsten Pauly et al.
PloS one, 3(7), e2681-e2681 (2008-07-17)
Functional and structural alterations of clustered postsynaptic ligand gated ion channels in neuronal cells are thought to contribute to synaptic plasticity and memory formation in the human brain. Here, we describe a novel molecular mechanism for structural alterations of NR1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service