Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3708

Sigma-Aldrich

Anti-Resistin Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$594.00

$594.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$594.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$594.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Retn(57264)

Specificity

Mouse resistin. Resistin is a protein produced by adipose tissue and released into the circulation that has been linked to insulin resistance and dietary obesity. The protein has been shown to be regulated by the anti-diabetic thiazolidinedione class of drugs.

Immunogen

Synthetic peptide from mouse resistin (aa 83-91).

Application

Detect Resistin using this Anti-Resistin Antibody validated for use in ELISA, WB & IC.
Immunoblotting: 1:500-1:1,000.

Immunocytochemistry: 1:150-1:300

ELISA: 1:20,000

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Physical form

Affinity purified immunoglobulin. Liquid in 20 mM PBS containing 0.1% sodium azide.
Format: Purified

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service