Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3272

Sigma-Aldrich

Anti-Aquaporin 1 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

antibody form

purified antibody

Quality Level

clone

polyclonal

species reactivity

rabbit, rat, human

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

General description

FUNCTION: SwissProt: P29972 # Forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.
SIZE: 269 amino acids; 28526 Da
SUBUNIT: Homotetramer.
SUBCELLULAR LOCATION: Membrane; Multi-pass membrane protein.
TISSUE SPECIFICITY: Expressed in a number of tissues including erythrocytes, renal tubules, retinal pigment epithelium, heart, lung, skeletal muscle, kidney and pancreas. Weakly expressed in brain, placenta and liver.
DOMAIN: SwissProt: P29972 Aquaporins contain two tandem repeats each containing three membrane-spanning domains and a pore-forming loop with the signature motif Asn-Pro-Ala (NPA).
SIMILARITY: Belongs to the MIP/aquaporin (TC 1.A.8) family.
MISCELLANEOUS: Pharmacologically inhibited by submillimolar concentrations of mercury.
Highly purified peptide corresponding to residues 242-260 of human Aquaporin 1 (SwissProt accession number P29972).

Specificity

Recognizes Aquaporin 1.

SPECIES REACTIVITIES: The epitope specific for Aquaporin 1 is identical in human, rodent, bovine and only slightly different in frog (18/19 aa). Reactivity with other species has not yet been tested.

Application

Western blotting: 1:500-1:1000 on rat kidney membranes.

Immunohistochemistry

Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 0.025% sodium azide. Reconstitute with 200 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). Reconstitute with 100 μL of deionized water. For negative control, preincubate 1.0 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.
The epitope specific for Aquaporin 1 is identical in human, rodent, bovine and sheep and only slightly different in canine (18/19 amino acids) and frog (14/19 amino acids).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service