Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AB2285

Sigma-Aldrich

Anti-Glutamate Receptor Delta 1/2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

GluR delta-1, glutamate receptor, ionotropic, delta 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$529.00

$529.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$529.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$529.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, primate, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRID1(2894)

General description

The majority of rapid excitatory transmission in the nervous system is mediated by glutamate-activated ion channels. Ionotropic glutamate receptors have been subdivided pharmacologically into AMPA (cx-amino-3-hydroxy-5methyl-Cisoxazole propionic acid), KA (kainate), and NMDA subtypes (Monaghan et al., 1989). These receptors have been implicated in a number of physiological and pathological processes. During development glutamate receptors are involved in neuronal differentiation, migration, and activity-dependent synapse formation (Balazs et al., 1989; Brewer, 1989; Komoru and Rakic, 1993). In mature neurons, long-term potentiation and long-term depression require ionotropic glutamate receptors. Glutamate-gated ion channels also play a central role in mediating the excitotoxic effects of glutamate (Olney, 1990). Pathophysiological processes thought to involve glutamate excitotoxicity and neurodegeneration include epilepsy stroke, Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and amyotrophic lateral sclerosis.

Specificity

Cat. # AB2285 reacts with the C-Terminus of Glutamate Receptor Delta 1/2.
Reacts weakly with mouse. May react with Primate based on sequence homology. Reactivity with other species has not been tested.

Immunogen

Epitope: C-Terminus, Cytoplasmic
Synthetic linear peptide to the C-Terminus, conjugated to BSA.

Application

Anti-Glutamate Receptor Delta 1/2 Antibody detects level of Glutamate Receptor Delta 1/2 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western Blotting:
Human Cerebellum Lysate.

Quality

Western Blotting: 1:5,000 dilution from a previous lot detected Glutamate Receptor Delta 1/2 on 10 µg of rat or human cerebellum lysate.

Target description

~114 kDa

Physical form

Antigen Affinity Purified
Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Rat cerebellum tissue lysate and human cerebellum tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Ren et al.
Neuroscience bulletin, 38(4), 342-358 (2021-12-16)
Central sensitization is essential in maintaining chronic pain induced by chronic pancreatitis (CP), but cortical modulation of painful CP remains elusive. Here, we examined the role of the anterior cingulate cortex (ACC) in the pathogenesis of abdominal hyperalgesia in a
Inés González-Calvo et al.
eLife, 10 (2021-03-05)
Fine control of protein stoichiometry at synapses underlies brain function and plasticity. How proteostasis is controlled independently for each type of synaptic protein in a synapse-specific and activity-dependent manner remains unclear. Here, we show that Susd4, a gene coding for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service