Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB2038

Sigma-Aldrich

Anti-Collagen Type V Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
radioimmunoassay: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Antibody shows less than 0.1% reactivity with rat Collagen Types I, II, III and rat Elastin by radioimmunoassay.

Immunogen

Collage Type V extracted and purified from rat skin.

Application

Anti-Collagen Type V Antibody is an antibody against Collagen Type V for use in ELISA, IF, RIA, IH(P).
Immunofluorescence: 1:40 dilution on frozen rat skin and liver tissues.

Immunohistochemistry: 1:250 on paraffin embedded rat tissues

Radioimmunoassay

ELISA

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneity in arterial remodeling among sublines of spontaneously hypertensive rats.
Bakker, EN; Groma, G; Spijkers, LJ; de Vos, J; van Weert, A; van Veen, H; Everts et al.
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service