Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB1962

Sigma-Aldrich

Anti-Paxillin Antibody, phospho-specific [Ser178]

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer178)

Gene Information

human ... PXN(5829)

General description

Paxillin, a focal adhesion protein, is involved in focal adhesion formation during cell adhesion and migration. Paxillin contains LD motifs, LIM domains, and SH3-/SH2-binding domains that participate in a variety of protein-protein interactions with kinases, GTPase-activating proteins, and cytoskeletal proteins. Phosphorylation of paxillin occurs at both tyrosine and serine sites. Serine phosphorylation of paxillin occurs in response to growth-factor activation and fibronectins. Both JNK1 and cdc2 kinases can phosphorylate serine 178 in paxillin. The mutant form of paxillin (S178A) decreases the migration of keratocytes and epithelial cells. Thus, phosphorylation paxillin at serine 178 may be important during cell migration.

Specificity

This antibody detects a 68 kDa protein corresponding to the molecular weight of phosphorylated paxillin on SDS-PAGE immunoblots of EGF treated A431 cells, but not in A431 control cells. Similar results were seen in calyculin A treated human A431 and aortic endothelial cells.

Immunogen

Epitope: phosphoserine 178
Synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 178 of human paxillin. This human sequence is highly conserved in rat and mouse paxillin.

Application

Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
This Anti-Paxillin Antibody, phospho-specific [Ser178] is validated for use in ELISA, WB for the detection of Paxillin.
Western blot: 1:500 ELISA: 1:2000

Physical form

Affinity-purified antibody is supplied in 100 μl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C for one year from date of receipt. Do not aliquot.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service