Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AB1868P

Sigma-Aldrich

Anti-Stromal Cell-Derived Factor-1α Antibody

Chemicon®, from rabbit

Synonym(s):

SDF-1alpha

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$446.00

$446.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$446.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CXCL12(6387)

Specificity

Reacts with human SCDF-1alpha

Immunogen

Highly pure (>98%) recombinant human SDF-1alpha.

Application

Detect Stromal Cell-Derived Factor-1α using this Anti-Stromal Cell-Derived Factor-1α Antibody validated for use in ELISA, WB, NEUT.
ELISA: 0.5 μg/mL this dilution will allow detection (in direct ELISA) of 0.2-0.4 ng/well when human SDF-1alpha is used to coat the plate.

Western blot: 0.1-0.2 μg/mL, this dilution should offer a detection limit of 1.5-3.0 ng/lane, under reducing or non-reducing conditions.

Optimal working dilutions must be determined by the end user.

Neutralization: To yield one-half maximal inhibition (ND50) of the biological activity of human SDF-1alpha (100 ng/mL), a concentration of 2.0-0.4 ng/well of recombinant human SDF-1alpha
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Physical form

Affinity purified immunoglobulin. Lyophilized from 0.5X PBS pH 7.2. Reconstitute with sterile distilled water.

Storage and Stability

The lyophilized antibody is stable at room temperature for at least one month and for greater than a year when kept at -20°C. When reconstituted in sterile water to a concentration of =0.2 mg/ml the antibody is stable for at least six weeks at 2-4°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service