Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB1665

Sigma-Aldrich

Anti-Fas Ligand Antibody

serum, Chemicon®

Synonym(s):

FasL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

should not react with

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FAS(355)

Immunogen

An 18 amino acid peptide sequence (aa 130-146) from rat FASL. This sequence is known to be 94% homologous with mouse FASL, differing by one amino acid. This peptide region does not show significant homology with human FASL, with only four amin

Application

Detect Fas Ligand using this Anti-Fas Ligand Antibody validated for use in ELISA & WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western blotting:1:1000 - 1:5,000, using Chemiluminescent detection. The use of 0.1 - 0.5% milk in diluents is recommended to reduce background. Based upon the calculated amino acid sequences, rat (278), mouse (279), and human (281) FASL has a molecular weight of ~32 kDa. A 40 kDa FASL band has been detected in mouse T cells.

ELISA: 1:10,000 - 1:50,000, for detection of control peptide (AG626) coated at a 1 μg/mL concentration.

Optimal working dilutions must be determined by end user.

Physical form

Liquid antiserum containing 0.05% sodium azide as a preservative.

Storage and Stability

Store at -20°C for up to 12 months in undiluted aliquots. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriella Brunlid et al.
Stem cells (Dayton, Ohio), 25(10), 2551-2558 (2007-07-07)
The potential of pluripotent embryonic stem (ES) cells to develop into functional cells or tissue provides an opportunity in the development of new therapies for many diseases including neurodegenerative disorders. The survival of implanted cells usually requires systemic immunosuppression, however

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service