Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB1597P

Sigma-Aldrich

Anti-Vesicular Monoamine Transporter 1 Antibody

Chemicon®, from rabbit

Synonym(s):

VMAT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Vesicular Monoamine Transporter 1 (VMAT1). An antibody made to the C-terminal VMAT1 peptide detected a major band at ~55 kDa in postnuclear supernatants of CHO transfected with VMAT1 and not in wild type cells (Liu et al. 1994). Some additional bands, both higher and lower molecular weight, were also detected and remain unaffected by the inclusion of protease inhibitors. VMAT1 has been localized in adrenal chromaffin cells, endocrine and paracrine cells associated with the intestine, stomach and sympathetic nervous system (Liu et al. 1994; Weihe et al. 1994). VMAT1 has also been found in endosomes of transfected CHO cells and in LDCVs of rat PC12 cells (Liu et al. 1994). This antibody is not expected to cross-react with human VMAT1 (Erickson et al. 1996).

Immunogen

A 12 amino acid C-terminal peptide sequence from rat VMAT1 (Liu et al. 1992). This peptide is predicted to be cytoplasmic.

Application

Anti-Vesicular Monoamine Transporter 1 Antibody detects level of Vesicular Monoamine Transporter 1 & has been published & validated for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-10 μg/mL (Chemiluminescence technique)

Immunohistochemistry: 1-20 μg/mL on paraformaldehyde fixed sections.

ELISA: 0.5-1.0 μg/mL (1 μg/mL VMAT1 peptide (Catalog Number AG250)/well)

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vesicular monoamine transporter 1 mediates dopamine secretion in rat proximal tubular cells.
Maurel, A; Spreux-Varoquaux, O; Amenta, F; Tayebati, SK; Tomassoni, D; Seguelas et al.
American Journal of Physiology: Renal Physiology null
Alex Ismail et al.
FEBS letters, 591(1), 137-144 (2016-12-08)
Rat perivascular adipose tissue (PVAT) stores, takes up, and releases norepinephrine (NE; Ayala-Lopez et al. (2014) Pharmacol Res Perspect 2, e00041). We hypothesized that 3T3-L1 adipocytes would exhibit similar behaviors and, thus, could serve as a model for PVAT adipocytes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service