Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB15500

Sigma-Aldrich

Anti-Calcium Transporter 1 Antibody

Chemicon®, from rabbit

Synonym(s):

CaT-1, ECAC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

CaT-1 (rat/mouse 727 aa; human 725 aa, chromosome 7q33-q34; ~85 kDa) is also known as ECAC2, CAT-like (CATL) or TRPV6 (transient receptor potential cation channel, subfamily V, member 6) contains a cytoplasmic N-terminus, 6 ankyrin repeat domains, 6 transmembrane domains, an extracellular pore region between TM5-6, and cytoplasmic C-terminus. CaT-1 shows 75% sequence identity to epithelial Ca++ channel (ECaC) protein from rabbit kidney. In rat, CaT-1 transcript (3-Kb) is expressed in duodenum, proximal jejunum cecum and colon but not in kidney; whereas ECaC in rabbit is expressed in apical cell membranes in distal part of nephron (kidney), duodenum, jejunum and placenta.

Specificity

Calcium transporter 1 (CaT-1, ECAC2). The immunogen sequence has no significant homology with CaT-2 or other proteins.
Other species have not yet been tested. The immunogen sequence is 100% conserved in mouse, 88% in humans and 94% in Cat-1/htrp8A and htrp8b proteins.

Immunogen

Synthetic peptide near the C-terminus of rat CaT-1.

Application

Anti-Calcium Transporter 1 Antibody detects level of Calcium Transporter 1 & has been published & validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-10 µg/mL using ECL. The expected size of CaT-1 is ~83 kDa. However, posttranslational modification may alter its size depending upon the species and cell type.
Immunohistochemistry: Not tested. It is recommended that the antibody be tried at 2-20 µg/mL
ELISA: 1:10,000-1:100,000 using 50-100 ng of control peptide/well.
Optimal working dilutions must be determined by end user.

Physical form

ImmunoAffinity Purified
Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service