Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB15334

Sigma-Aldrich

Anti-Nab2 Antibody

serum, Chemicon®

Synonym(s):

NGFI-A-binding protein-2, EGR-1-binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NAB2(4665)

Specificity

Nab2 (NGFI-A-binding protein-2, EGR-1-binding protein 2). By Western blot the antibody recognizes a band at ~57 kDa in mouse brain lysate.

Application

Anti-Nab2 Antibody detects level of Nab2 & has been published & validated for use in WB.
Research Sub Category
Neuroinflammation & Pain

Developmental Neuroscience
Western blot: 1:1,000-1:5,000 on extracts from mouse brain lysate.

Optimal working dilutions must be determined by the end user.

Target description

57 kDa

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samira Chandwani et al.
PloS one, 8(10), e76842-e76842 (2013-11-10)
Neurotrophins and modifiers of chromatin acetylation and deacetylation participate in regulation of transcription during neuronal maturation and maintenance. The striatal medium spiny neuron is supported by cortically-derived brain derived neurotrophic factor and is the most vulnerable neuron in Huntington's disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service