Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB15188

Sigma-Aldrich

Anti-Neurofascin Antibody, common epitope

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NFASC(23114)

Specificity

Neurofascin, common epitope. By Western blot the antibody recognizes a band at ~186 kDa in mouse brain lysate.

Immunogen

Synthetic peptide from the COOH terminus of the intracellular domain common to both NF186 and NF155.

Application

Anti-Neurofascin Antibody, common epitope is an antibody against Neurofascin for use in WB.
Research Category
Neuroscience
Research Sub Category
Neural Stem Cell Migration

Developmental Neuroscience
Western blot: 0.5-1.0 μg/mL on mouse brain lysate.

Optimal working dilutions must be determined by end user.

Target description

~186 kDa

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% sodium azide.
Format: Purified
ImmunoAffinity Purified

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michell M Reimer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(49), 18185-18194 (2011-12-14)
Myelinated axons have a distinct protein architecture essential for action potential propagation, neuronal communication, and maintaining cognitive function. Damage to myelinated axons, associated with cerebral hypoperfusion, contributes to age-related cognitive decline. We sought to determine early alterations in the protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service