Skip to Content
MilliporeSigma
All Photos(4)

Documents

AB15138

Sigma-Aldrich

Anti-Nogo Receptor Antibody

from rabbit

Synonym(s):

Reticulon-4 receptor, Nogo receptor, NgR, Nogo-66 receptor, Nogor, Rtn4r

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rtn4R(65079)

General description

Axons are essential for neuronal communication but they do not regenerate after injury to the adult mammalian brain or spinal cord. Failed regeneration is due in part to the production of a potent axonal growth inhibitor, Nogo, by myelinating cells. The finding of a high affinity axonal receptor for the extracellular domain of Nogo provides the first insight into the basis of Nogo action. Disrupting the interaction of Nogo with the Nogo-66 receptor may facilitate axonal regeneration in vivo. The protein is dubbed the Nogo receptor because it binds with several other proteins that block neural growth. It is found to be ubiquitous in the brain and spinal cord.

Specificity

Cat. # AB15138 recognizes the mature form of Nogo Receptor.
May react with human and primate based on sequence homology. Reactivity with other species has not been determined.

Immunogen

Epitope: Mature Form
Recombinant protein.

Application

Anti-Nogo Receptor Antibody detects level of Nogo Receptor & has been published & validated for use in WB, IH.
Non-Lot Specific Tested Application 1:
Immunohistochemistry: Working dilutions of 1:50-1:500 in rat brain and spinal cord (paraffin embedded)
Research Category
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance

Quality

Routinely tested on mouse cerebellum lysate.
Lot Specific Tested Application 1: Western Blot

Target description

~50.9 kDa

Physical form

Format: Purified
Protein A purified
Purified in PBS with 0.1% NaN3

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
Mouse cerebellum lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Tao Liu et al.
Experimental and therapeutic medicine, 22(6), 1413-1413 (2021-10-23)
Methylprednisolone (MP) is widely used to treat clinical spinal cord injury (SCI). Treadmill training is also considered an important treatment after SCI to improve motor function in patients, resulting in an evident improvement. Therefore, the present study was designed to
Hongyan An et al.
Journal of cell science, 129(6), 1198-1209 (2016-01-31)
Inhibitory proteins, particularly Nogo 66, a highly conserved 66-amino-acid loop of Nogo A (an isoform of RTN4), play key roles in limiting the intrinsic capacity of the central nervous system (CNS) to regenerate after injury. Ligation of surface Nogo receptors
Daniela Talhada et al.
International journal of molecular sciences, 22(19) (2021-10-14)
Dopaminergic treatment in combination with rehabilitative training enhances long-term recovery after stroke. However, the underlying mechanisms on structural plasticity are unknown. Here, we show an increased dopaminergic innervation of the ischemic territory during the first week after stroke induced in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service