Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB15080

Sigma-Aldrich

Anti-Frataxin Antibody

from rabbit

Synonym(s):

Anti-CyaY, Anti-FA, Anti-FARR, Anti-FRDA, Anti-X25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FXN(2395)
mouse ... Fxn(14297)
rat ... Fxn(499335)

General description

Frataxin is a nuclear encoded protein that is found in the mitochondria. This protein is not well understood but may be involved with iron sulfur clusters. In humans the gene is localized on chromosome 9 and is highly conserved during evolution. The gene is expressed in every cell, although in varying levels in different tissues and during development. The specific function of Frataxin is still unknown, but it has been shown to play a role in iron metabolism. Studies have demonstrated that the deletion of the Frataxin gene in yeast results in iron accumulation in mitochondria and loss of respiration. Recombinant human Frataxin has been shown to bind iron in vitro, and increased mitochondrial iron levels have been observed in patients with Friedreich′s Ataxia (FRDA).

Specificity

Cat. # AB15080 recognizes Frataxin.

Immunogen

Epitope: mature chain
GST-tagged Frataxin corresponding to mature chain.

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-Frataxin Antibody is validated for use in WB for the detection of Frataxin.

Quality

Routinely evaluated by Western Blot on Human heart lysates.

Western Blot Analysis: 1:1,000 - 1:10,000 dilution of this lot detected Frataxin on 10 ug of human heart lysate

Target description

Approx. 18 kDa

Physical form

Format: Purified
GST-depleted sera
Rabbit polyclonal serum in buffer containing 0.05% NaN3.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Human heart lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service