Skip to Content
MilliporeSigma
All Photos(1)

Documents

AB10418

Sigma-Aldrich

Anti-Puma BH3 domain Antibody

from rabbit

Synonym(s):

BCL2 binding component 3, Bcl-2 binding component 3, p53 up-regulated modulator of apoptosis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Bbc3(170770)

General description

PUMA is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMA-a and PUMA-b. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis

Specificity

Other species have not been tested.
The antibody reacts with the 21 kDa Puma BH3 domain protein.

Immunogen

This antibody was developed against a synthetic peptide selected from the C-terminal region of human Puma.

Application

Anti-Puma BH3 domain Antibody is an antibody against Puma BH3 domain for use in WB.

Quality

Western Blot Analysis: 1:250 dilution of this antibody detected Puma BH3 domain on mouse brain lysate.

Target description

21 kDa

Physical form

Format: Purified

Analysis Note

Control
Western Blot : Mouse brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhargab Kalita et al.
PloS one, 11(12), e0168525-e0168525 (2016-12-31)
Development of an effective radio protector to minimise radiation-inflicted damages have largely failed owing to inherent toxicity of most of the agents examined so far. This study is centred towards delivering protection to lethally irradiated mice by pre-administration of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service