Skip to Content
MilliporeSigma
All Photos(2)

Documents

AAWP03700

Millipore

MCE Membrane Filter, 0.8 μm Pore Size

MF-Millipore, filter diam. 37 mm, hydrophilic, white, for aerosol monitoring

Synonym(s):

MF-Millipore Membrane Filter, 0.8 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

cellulose membrane
mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

Agency

in accordance with ASTM® D2914,D4185,D4765
in accordance with NIOSH 5515,6016,7020,7024,7027,7029,7030,7048,7056,7082,7102,7105,7300,7900,7902

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

16 L/min-cm2 air flow rate
190 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

37 mm

thickness

150 μm

color

white

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.8 μm pore size
82 % porosity

bubble point

≥1.0 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: AAWP

Application

  • Air monitoring
  • Particle monitoring
  • Particle removal
  • Bioassays
  • Microplastics

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service