Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

957571

Supelco

Dosing aid made of PP, 20ml, 28mm, natural for Extran®, AWH

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$18.00

$18.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$18.00

About This Item

UNSPSC Code:
41122409
NACRES:
NB.21

$18.00


Please contact Customer Service for Availability

Request a Bulk Order

Packaging

Fibre carton

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service