Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

9310-OP

Millipore

TRIS Hydrochloride

OmniPur® Grade, >=99.0%

Synonym(s):

Trizma® hydrochloride, Trizma, Tris(hydroxymethyl)aminomethane, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$244.00
5 KG
$1,600.00

$244.00


Available to ship onApril 02, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$244.00
5 KG
$1,600.00

About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

$244.00


Available to ship onApril 02, 2025Details


Request a Bulk Order

Quality Level

form

crystalline powder
needles

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

useful pH range

7.0-9.0

pKa (25 °C)

8.1

shipped in

ambient

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Biology Grade.
260 (10%, water): ≤0.05 AU
A280 (10%, water): ≤0.05 AU
Assay: ≥99%
DNAse: none detected
Identity IR spectrum: conforms
Lead (Pb): ≤.0002%
Phosphatase: none detected
Protease: none detected
RNAse: none detected
Water (H2O): ≤0.5%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Use of tetraethylammonium (TEA) and Tris loading for blocking TRPM7 channels in intact cells.: This study highlights the application of TRIS Hydrochloride in blocking TRPM7 channels, which is significant for cell signaling and ion channel regulation in pharmacological research (Holderby & Kozak, 2024).

  • Investigating the influence of solvent type and pH on protein adsorption onto silica surface by evanescent-wave cavity ring-down spectroscopy.: This study uses TRIS Hydrochloride to explore protein-silica interactions, providing insights into surface chemistry and biomolecular adsorption processes (Alnaanah & Mendes, 2024).

Warning

Toxicity: Irritant (B)

Other Notes

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service