Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

9011S

Sigma-Aldrich

Human Specific–Ultra Sensitive Insulin

Sign Into View Organizational & Contract Pricing

Select a Size

250 TUBES
$158.00

$158.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 TUBES
$158.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84
form:
lyophilized

$158.00


Please contact Customer Service for Availability

Request a Bulk Order

conjugate

125Iodine

Quality Level

label

125Iodine labeled

form

lyophilized

NCBI accession no.

UniProt accession no.

General description

Radioactivity: <3 μCi
These iodinated hormonos are HPLC purified and prepared fresh monthly.

Application

Research Category
All
appropriate label hydrating buffer if kit replacement or assay buffer of choice

Storage and Stability

2–8 °C

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service