Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8920-OP

Millipore

TEMED

OmniPur® Grade

Synonym(s):

OmniPur® TEMED, Tetramethylethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
41105319

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear

shipped in

ambient

InChI

1S/C6H16N2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3

InChI key

KWYHDKDOAIKMQN-UHFFFAOYSA-N

General description

Free radical initiator and stabilizer used in acrylamide polymerization for PAGE gels. Molecular Biology Grade.

DNAse: none detected
Residue after ignition: ≤0.005%
Moisture (H2O): ≤0.5%
Protease: none detected
Purity (anhydrous basis): ≤99.0%
RNAse: none detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Flammable (J)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.7 °F - closed cup

Flash Point(C)

16.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service