Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

869003-M

Sigma-Aldrich

Luteinizing Hormone, Human Pituitary, Iodination Grade

Synonym(s):

Luteinizing Hormone, Human Pituitary, Iodination Grade, hLH

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$195.00
50 μG
$672.00

$195.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$195.00
50 μG
$672.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
biological source:
human
form:
solid

$195.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

description

Merck USA index - 14, 5613

Assay

≥95% (weight)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.05% hCG
≤0.5% hFSH, hTSH

color

white

solubility

water: 1 mg/mL

storage temp.

−20°C

General description

Native, human luteinizing hormone from pituitary gland. Heterodimeric glycoprotein hormone secreted by the anterior pituitary gland that is required for ovulation, spermatogenesis, and the biosynthesis of sex steroids. Regulates biosynthesis and/or secretion of sex steroids. In the female, LH stimulates synthesis of progesterone in the ovaries, release of estrogen from Graafian follicles, induction of ovulation, and formation of the corpus luteum. In the male, LH stimulates testosterone secretion by the testes. Biological specificity has been attributed to the β-chain.

Biochem/physiol Actions

≥ 5,000 IU/mg

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Other Notes

Ramanujam, L.N., et al. 1999. Clin. Endocrinol.51, 243.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Could you please provide instructions on how to reconstitute product number 869003, specifically Luteinizing Hormone, Human Pituitary, Iodination Grade?

    1 answer
    1. The solubility of gonadorelin (acetate) in ethanol is roughly 0.25 mg/ml and about 30 mg/ml in DMSO and DMF.

      Helpful?

  2. Hello, Can this iodination grade hLH be used in rat LH radioimmunoassay or do you carry any iodination grade rLH?

    1 answer
    1. The suitability of this product in rat studies has not been determined. It has been shown that iodination may lower the potency of LH and a lower affinity to hormonal receptors. It may be helpful to view a similar product L6420. Again, this product has not been tested for suitability in rat.


      Below are some references that may be helpful in determining if these products would be suitable for the specified application.

      1. Effect of Iodination on the Biological Activity and Metabolism of Human Luteinizing Hormone
      https://academic.oup.com/jcem/article-abstract/28/5/613/2715632

      2. Iodination of Luteinizing Hormone-Releasing Hormone
      https://pubs.acs.org/doi/pdf/10.1021/bi00553a006

      3. Species specificity in the response of an in vitro amphibian (Xenopus laevis) ovulation assay to mammalian luteinizing hormones
      https://www.sciencedirect.com/science/article/pii/0016648076900393

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service