Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.57014

Sigma-Aldrich

Deblocking Reagent B for DNA Synthesis

2% Trichloroacetic Acid in Dichloromethane (w,w), Novabiochem®

Synonym(s):

2% Trichloroacetic Acid in Dichloromethane (w/w), Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
$174.00
450 ML
$197.00

$174.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 L
$174.00
450 ML
$197.00

About This Item

UNSPSC Code:
41116006
NACRES:
NA.21

$174.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

reaction suitability

reaction type: Acetylations
reagent type: Lewis acid

General description

Pre-mixed ready-to-use Oxidizer Reagent
  • 857014.0450: Compatible with MerMade DNA Synthesisers. Attaches directly to Instrument (28/405 neck finish)
  • 857014.4000: Compatible with MerMade DNA Synthesisers. Stand alone from Instrument (38mm Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service