Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.56189

Sigma-Aldrich

H-Pro-Trityl-NovaPEG resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641

Quality Level

form

beads

manufacturer/tradename

Novabiochem®

storage temp.

2-8°C

Analysis Note

Colour (visual): slight yellow to beige to orange
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.15 - 0.40 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service