Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.56146

Sigma-Aldrich

Fmoc-Asp(Wang-resin)-AMC (100-200 mesh)

Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$640.00
1 G
$1,070.00

$640.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$640.00
1 G
$1,070.00

About This Item

UNSPSC Code:
12352209
NACRES:
NA.22

$640.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

General description

A novel resin for the preparation of peptide substrates based on 3-amino-7-methylcoumarin (AMC) by Fmoc SPPS. Following Fmoc removal with 20% piperidine, peptide assembly can be effected using standard coupling methods. Treatment with 95% TFA releases the peptide-AMC directly from the solid phase.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Linkage

Replaces: 04-12-3915

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 1.00 mmol/g
Enantiomeric purity: lot specific result
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene-1% DVB), 200 - 400 mesh. The resin is endcapped after coupling the amino acid derivative.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service