Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.55136

Sigma-Aldrich

PAL-NovaPEG resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

product line

NovaPEG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

PAL-NovaPEG resin consists of Barany′s aminomethyl-dimethoxyphenoxyvaleric acid linker [1] attached to NovaPEG amino resin. The amino group of this linker can be easily acylated under standard coupling conditions. Following peptide assembly, treatment with 95% TFA containing scavengers releases the desired peptide amide. Studies have shown the acid sensitivity of this linker to be around twice that of the Rink amide linker [2].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to yellow to beige/brown to orange
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.30 - 0.70 mmol/g
Swelling Volume (in DMF): lot specific result

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service