Skip to Content
MilliporeSigma
All Photos(2)

Documents

8.55064

Sigma-Aldrich

NovaSyn® TG hydroxy resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
13111023
NACRES:
NA.22

Quality Level

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-30°C

General description

NovaSynTG hydroxy resin is a composite of low cross-linked polystyrene and 3000-4000 M.W. polyethylene glycol, in which the ends of the PEG chains have been functionalized with hydroxyl groups. The 130 μm beads have a narrow size distribution, high diffusion rates and excellent swelling across a wide range of solvents from toluene to water [1,2,3], making them ideally suited to the synthesis of serial and parallel libraries by organic solid phase synthesis. This support has been used for the solid phase immobilization of carboxylic acids [4,5,6,7,8,9], with cleavage by saponification with dilute aq. NaOH.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] W. Rapp, et al. in ′Innovation & Perspectives in Solid Phase Synthesis, 1st International Symposium′, R. Epton (Eds), SPCC UK Ltd., Birmingham, 1990, pp. 205.
[2] L. Zhang, et al. in ′Peptides 1990, Proc. 21st European Peptide Symposium′, E. Giralt & D. Andreu (Eds), ESCOM, Leiden, 1990, pp.196.
[3] E. Bayer (1991) Angew. Chem. Int. Ed. Engl., 30, 113.
[4] M. Hiroshige, et al. (1995) Tetrahedron Lett., 36, 4567.
[5] M. C. Fagnola, et al. (1997) Tetrahedron Lett., 38, 2307.
[6] D. Fancelli, et al. (1997) Tetrahedron Lett., 38, 2311.
[7] M. Patek, et al. (1995) Tetrahedron Lett., 36, 2227.
[8] V. Krchnak, et al. (1995) Tetrahedron Lett., 36, 6193.
[9] A. K. Szardenings & T. S. Burkoth (1997)Tetrahedron, 53, 6573.

Linkage

Replaces: 01-64-0096

Analysis Note

Color (visual): white to amber to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.20 - 0.30 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
130 µm beads

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service