Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.53019

Sigma-Aldrich

Boc-Lys(Fmoc)-OH

≥99.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Boc-Lys(Fmoc)-OH, N-α-t.-Boc-N-ε-Fmoc-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N2O6
CAS Number:
Molecular Weight:
468.54
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

product name

Boc-Lys(Fmoc)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)
≥98.0% (acidimetric)
≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

InChI

1S/C26H32N2O6/c1-26(2,3)34-25(32)28-22(23(29)30)14-8-9-15-27-24(31)33-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H,27,31)(H,28,32)(H,29,30)

InChI key

JYEVQYFWINBXJU-UHFFFAOYSA-N

General description

Building block for preparation of peptides modified at lysine side chain by Boc SPPS. Fmoc group can be removed with piperidine in DMF.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Linkage

Replaces: 04-12-0069

Analysis Note

Color (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Purity (HPLC): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): passes test
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 1.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service