Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52412

Sigma-Aldrich

Fmoc-His(MBom)-OH

N-α-Fmoc-3-methoxybenzyloxymethyl-L-histidine Novabiochem®

Synonym(s):

Fmoc-His(MBom)-OH, N-α-Fmoc-N-π-(p-methoxybenzyloxymethyl)-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H29N3O6
Molecular Weight:
527.57
UNSPSC Code:
12352209

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

General description

A novel histidine derivative for Fmoc SPPS in which the π-imidazole nitrogen is blocked with the acid-labile MBom group, In contrast to Fmoc-His(Trt)-OH, coupling of this derivative is free from the risk of racemization, even under base-mediated coupling conditions. Methoxyamine should be added to the cleavage mixture to scavenge liberated formaldehyde.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] H. Hibino et al. (2011) Tetrahedron lett., 52, 4947.

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Purity (HPLC): ≥ 98.0 % (a/a)
Purity (DC(BAWO)): ≥ 98 %
Identity (IR): passes test
Solubility (1 mmole in 2 ml DMF): clear soluble
Enantiomeric purity: ≥ 99.5 % (a/a)
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service