Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.52378

Sigma-Aldrich

Fmoc-Phe(SO3Na)-OH

Novabiochem®

Synonym(s):

Fmoc-Phe(SO3Na)-OH, N-α-Fmoc-4-sulfophenylalanine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20NNaO7S
Molecular Weight:
489.47
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

sulfonic acid

storage temp.

15-25°C

General description

Introduction of 4-sulfo-phenylalanine is an effective strategy for increasing solubility of a peptide. This amino acid has also been employed in analogs of angiotensin 2 [1], phytosulfokine [2], and cyclolinopeptide A [3]. Its incorporation during solid phase synthesis using Fmoc-Phe(SO3Na)-OH is best carried out with base-mediated coupling methods such as PyBOP® or HBTU.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] E. Escher, et al. (1983) , 66, 1355.
[2] A. Bahyrycz, et al. (2004) , 10, 462.
[3] Y. Fujiwara, et al. (1994) , 42, 724.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in DMF): -17.0 - -15.0 °
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Purity TLC (BAW 1): ≥ 97.0 %
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service