Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52371

Sigma-Aldrich

Fmoc-His(Clt)-OH

for peptide synthesis, Novabiochem

Synonym(s):

Fmoc-His(Clt)-OH, N-α-Fmoc-N-im-2-chlorotrityl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H32ClN3O4
CAS Number:
Molecular Weight:
654.15
UNSPSC Code:
12352209
Pricing and availability is not currently available.

Product Name

Fmoc-His(Clt)-OH, Novabiochem®

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

InChI

1S/C40H32ClN3O4/c41-36-22-12-11-21-35(36)40(27-13-3-1-4-14-27,28-15-5-2-6-16-28)44-24-29(42-26-44)23-37(38(45)46)43-39(47)48-25-34-32-19-9-7-17-30(32)31-18-8-10-20-33(31)34/h1-22,24,26,34,37H,23,25H2,(H,43,47)(H,45,46)/t37-/m0/s1

InChI key

SYGADBVEKBWUKO-QNGWXLTQSA-N

General description

A useful derivative for the convergent synthesis of peptide due to the enhanced stability of Clt group compared to Trt group to mildy acidic reagents. Fmoc-His-(Clt)-OH gives approximately a third less racemization when compared to Fmoc-His(Trt)-OH during slow coupling reactions.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service