Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52352

Sigma-Aldrich

(2S,3S)-Fmoc-Abu(3-N3)-OH

Novabiochem®

Synonym(s):

(2S,3S)-Fmoc-Abu(3-N3)-OH, (2S,3S)-2-(Fmoc-amino)-3-azidobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N4O4
CAS Number:
Molecular Weight:
366.37
UNSPSC Code:
12352209

Quality Level

product line

Novabiochem®

Assay

≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

azide

storage temp.

15-25°C

General description

A useful tool for the synthesis of branched, side-chain modified, cyclic peptides and peptide tools for click chemistry. The side-chain azido group is completely stable to piperidine and TFA, but can be readily converted to an amine on the solid phase or in solution by reduction with thiols [1] or phosphines [2,3].

Associated Protocols and Technical Articles
Guide to Selection of Orthogonally-Protected Building Blocks for Fmoc SPPS

Literature references

[1] M. Meldal, et al. (1997) Tetrahedron Lett., 38, 2531.
[2] J. T. Lundquist & J. C. Pelletier (2001) Org. Lett.,3, 781.
[3] N. Nepomniaschiy, et al. (2008) Org.Lett., 10, 5243.

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service